Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Extension Cables

Extension Cables

PVC extension cable, White, 10 m
Catalog # 139-1302
Supplier:   BACHMANN ELEKTRO
Extension Cables
Catalog # 139-1302
Supplier:  BACHMANN ELEKTRO
Extension Cables
Extension Cables

PVC extension cable, White, 10 m

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

PVC extension cord with moulded, double-earthed plug and Schuko coupling.

  • Cable (HO5VV-F G31,50 mm²) available in different lengths
  • Colour: white
  • Max. power: 10/16 A, 250 V~

Certifications: VDE compliant

Specifications
  • Colour:
    White
  • Description:
    PVC extension cable
  • Length:
    10 m