Order Entry
Netherlands
ContactUsLinkComponent
Strip plate F8 polystyrene clear universal binding breakable F-bottom 1 * 250 items
Catalog # 735-0552
Strip plate F8 polystyrene clear universal binding breakable F-bottom 1 * 250 items
Catalog # 735-0552
Frequently Bought Together