Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Borane dimethylamine complex ≥97%

Borane dimethylamine complex ≥97%

Glass bottle for solids, 100 g
Catalog # 89149.22
Toxic
Corrosive
Flammable
Borane dimethylamine complex ≥97%
Catalog # 89149.22
CAS Number:  74-94-2
Borane dimethylamine complex ≥97%
Borane dimethylamine complex ≥97%

Glass bottle for solids, 100 g

CAS Number:  74-94-2
Price
211,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 74-94-2 Molecular Formula: BH3.NH(CH3)2 Formula Weight: 58.92 Physical Form: Crystalline Density: 0.69 Appearance: White Melting Point: 36° Boiling Point: 59-65°/1-2mm UN No.: 2925 MDL No.: MFCD00051068

Synonyms:  Boron hydride dimethylamine, Borane, dimethylamine complex
  • CAS Number:74-94-2
  • Formula:C₂H₁₀BN
  • MDL Number:MFCD00051068
  • Molecular Weight:58.92 g/mol
  • Boiling Pt:59…65 °C (1…2 mmHg)
  • Melting Pt:36 °C
  • Density:0.69
  • Storage Temperature:Ambient
  • EINECS:200-823-7
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications