Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Borane dimethylamine complex ≥97%

Borane dimethylamine complex ≥97%

Glass bottle for solids, 25 g
Catalog # 89149.14
Toxic
Corrosive
Flammable
Borane dimethylamine complex ≥97%
Catalog # 89149.14
CAS Number:  74-94-2
Borane dimethylamine complex ≥97%
Borane dimethylamine complex ≥97%

Glass bottle for solids, 25 g

CAS Number:  74-94-2
Price
69,90 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 74-94-2 Molecular Formula: BH3.NH(CH3)2 Formula Weight: 58.92 Physical Form: Crystalline Density: 0.69 Appearance: White Melting Point: 36° Boiling Point: 59-65°/1-2mm UN No.: 2925 MDL No.: MFCD00051068

Synonyms:  Boron hydride dimethylamine, Borane, dimethylamine complex
  • CAS Number:74-94-2
  • Formula:C₂H₁₀BN
  • MDL Number:MFCD00051068
  • Molecular Weight:58.92 g/mol
  • Boiling Pt:59…65 °C (1…2 mmHg)
  • Melting Pt:36 °C
  • Density:0.69
  • Storage Temperature:Ambient
  • EINECS:200-823-7
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.