Sodium hydroxide 30% (w/w) in aqueous solution
Plastic bottle, 100 mL
Leverancier: Thermo Fisher Scientific
Sodium hydroxide 30% (w/w) in aqueous solution
Catalogus # 87864.AE
Leverancier: Thermo Fisher Scientific
CAS-nummer: 1310-73-2
Ga naar:
Product Details & Documents
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Appearance: Colourless UN No.: 1824 MDL No.: MFCD00003548
Synoniemen: Caustic soda
- CAS-nummer:1310-73-2
- Formule:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Kokend Pt:119…135 °C (1013 hPa)
- Smelten Pt:9…10 °C
- Dichtheid:1,53 g/cm³ (20 °C)
- Temperatuur bij opslag:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Specificaties
Specificaties
- Pack type:Plastic bottle
- Pk:100 mL
Specificaties
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



