Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Yttrium(III) fluoride ≥98%

Glass bottle for solids, 25 g
Catalogus # 83106.14
Giftig
Irriterend
Leverancier:   Thermo Fisher Scientific
Yttrium(III) fluoride ≥98%
Catalogus # 83106.14
CAS-nummer:  13709-49-4
Yttrium(III) fluoride ≥98%
Yttrium(III) fluoride ≥98%

Glass bottle for solids, 25 g

CAS-nummer:  13709-49-4
Price
82,50 €
Log in voor beschikbaarheid
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 13709-49-4 Molecular Formula: F3Y Formula Weight: 145.90 Physical Form: -325 Mesh Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 1387° UN No.: 3288 MDL No.: MFCD00049714

Synoniemen:  Yttrium trifluoride, Yttrium(III) fluoride
  • CAS-nummer:13709-49-4
  • Formule:F₃Y
  • MDL-nummer:MFCD00049714
  • Molecular Weight:145.9 g/mol
  • Kokend Pt:2227 °C (1013 hPa)
  • Smelten Pt:1387 °C
  • Dichtheid:4.01 g/cm³ (20 °C)
  • Temperatuur bij opslag:Ambient
  • EINECS:237-257-5
  • UN:3288
  • ADR:6.1,III
  •  :Density: 4.01 g/cm³ (20 °C)
Specificaties

Specificaties

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specificaties