Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Glass Stoppers with Octagonal Grip
Glass Stoppers with Octagonal Grip
34/35, Amber glass
Catalog # 217-9152
Supplier:   witeg Labortechnik
Glass Stoppers with Octagonal Grip
Catalog # 217-9152
Supplier:  witeg Labortechnik
Glass Stoppers with Octagonal Grip
Glass Stoppers with Octagonal Grip
34/35, Amber glass
Price
15,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Solid standard taper stoppers with octagonal grip, made of Borosilicate glass 3.3.
  • Acc. to DIN 12252
* Semi-hollow
Specifications
  • NS:
    34/35
  • Type:
    Amber glass
  • Seq:
    05