Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Calcium hydride ≥92%

100 g
Catalog # 41590.22
Flammable
Calcium hydride ≥92%
Catalog # 41590.22
CAS Number:  7789-78-8
Calcium hydride ≥92%
Calcium hydride ≥92%

100 g

CAS Number:  7789-78-8
Price
138,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 92% min CAS No.: 7789-78-8 Molecular Formula: CaH2 Formula Weight: 42.10 Physical Form: 1-20mm Granules Density: 1.900 Melting Point: 600° dec. Solubility: Decomposes in water, alcohol and acids to H2 Merck Reference: 14,1672 UN No.: 1404 MDL No.: MFCD00010897

Synonyms:  Calcium dihydride
  • CAS Number:7789-78-8
  • Formula:H₂Ca
  • MDL Number:MFCD00010897
  • Molecular Weight:42.09 g/mol
  • Merck Index:14,1672
  • Melting Pt:600 °C (dec.)
  • Density:1.900
  • Storage Temperature:Ambient
  • EINECS:232-189-2
  • UN:1404
  • ADR:4.3,I
Specifications

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.