Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Morpholine ≥99.0% ACS

Glass bottle for solids, 500 g
Catalog # 31984.36
Corrosive
Flammable
Irritant
Morpholine ≥99.0% ACS
Catalog # 31984.36
CAS Number:  110-91-8
Morpholine ≥99.0% ACS
Morpholine ≥99.0% ACS

Glass bottle for solids, 500 g

CAS Number:  110-91-8
Price
58,60 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99.0% min CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972

  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:14,6277
  • Boiling Pt:129 °C
  • Melting Pt:–5 °C
  • Density:1.007
  • Flash Pt:35 °C (95 °F)
  • Storage Temperature:Ambient
  • EINECS:203-815-1
  • UN:2054
  • ADR:8,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications