Order Entry
Netherlands
Orders LinkContactUsLinkComponent
n-Dodecane ≥99%

n-Dodecane ≥99%

Glass bottle for solids, 100 g
Catalog # 31324.22
Health
n-Dodecane ≥99%
Catalog # 31324.22
CAS Number:  112-40-3
n-Dodecane ≥99%
n-Dodecane ≥99%

Glass bottle for solids, 100 g

CAS Number:  112-40-3
Price
56,70 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 112-40-3 Molecular Formula: C12H26 Formula Weight: 170.34 Flash Point: 71°(159°F) Physical Form: Liquid Density: 0.749 Refractive Index: 1.4220 Appearance: Colourless Odour: Petrol-like Melting Point: -10° Boiling Point: 215-217° Beilstein Reference: 1697175 MDL No.: MFCD00008969

Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:H₃C(CH₂)₁₀CH₃
  • MDL Number:MFCD00008969
  • Molecular Weight:170.34 g/mol
  • Boiling Pt:215…217 °C
  • Melting Pt:–10 °C
  • Density:0.749
  • Flash Pt:71 °C (159 °F)
  • Storage Temperature:Ambient
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.