Deep Well Platte, 2.2 ml, 96 Vertiefunge aus PP, U-Boden, viereckig, Beut/4 Stk TREFF (32 Stk) 32 Stk 1 * 32 ST
Catalog # TREF96.9799.9.01
Supplier: Nolato Treff AG
Deep Well Platte, 2.2 ml, 96 Vertiefunge aus PP, U-Boden, viereckig, Beut/4 Stk TREFF (32 Stk) 32 Stk 1 * 32 ST
Catalog # TREF96.9799.9.01
Supplier: Nolato Treff AG
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection