Order Entry
Netherlands
ContactUsLinkComponent
GenePhor Electrophoresis Unit (Order precast plastic backed gels and silver stain detection, buffers and electrode wicks separately) 1 * 1 items
Catalog # 18-1115-82
Supplier:  Cytiva
GenePhor Electrophoresis Unit (Order precast plastic backed gels and silver stain detection, buffers and electrode wicks separately) 1 * 1 items
Catalog # 18-1115-82
Supplier:  Cytiva
Frequently Bought Together