Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Accessories for transilluminators

Accessories for transilluminators

Replacement longwave (365 nm) UV tube, 8 W
Catalog # 730-1428
Accessories for transilluminators
Catalog # 730-1428
Accessories for transilluminators

Replacement longwave (365 nm) UV tube, 8 W

Price
26,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Accessories information: For information about any replacement tubes that are not listed, please contact your local Avantor office.

Specifications
  • Description:
    Replacement longwave (365 nm) UV tube, 8 W
  • Seq:
    03