Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Clamps
Clamps
20 - 40 mm
Catalog # 241-7315
Supplier:   USBECK Laborgeräte
Clamps
Catalog # 241-7315
Clamps
Clamps
20 - 40 mm
Price
13,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Zinc die-cast, powder coated.
  • The jaws of the clamps are covered with layer of cork to protect the clamped items
  • Without bosshead for fixing to stand
  • With wing screws for rapid easy clamping
Specifications
  • Spanning width:
    20 - 40 mm