Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Morpholine for synthesis, Sigma-Aldrich®

Plastic bottle, 100 mL
Catalogus # 8.06127.0100
Ontvlambaar
Giftig
Corrosief
Leverancier:   Merck
Morpholine for synthesis, Sigma-Aldrich®
Catalogus # 8.06127.0100
Leverancier:  Merck
CAS-nummer:  110-91-8
Morpholine for synthesis, Sigma-Aldrich®
Morpholine for synthesis, Sigma-Aldrich®

Plastic bottle, 100 mL

CAS-nummer:  110-91-8
Price
32,00 €
Log in voor beschikbaarheid
1 * 0,1 L
Product Details & Documents
  • CAS-nummer:110-91-8
  • Formule:C₄H₉NO
  • MDL-nummer:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck index:12,06362
  • Kokend Pt:128.9 °C (1013 hPa)
  • Smelten Pt:–5 °C
  • Dichtheid:1 g/cm³ (20 °C)
  • Flits Pt:38 °C
  • Temperatuur bij opslag:Ambient
  • UN:2054
  • ADR:8 (3),I
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-815-1
Specificaties

Specificatie testresultaten

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    87.12 g/mol
  • Boiling Point:
    128 - 130 °C (1013 hPa)
  • Vapor Pressure:
    10 hPa (20 °C)
  • Flash Point:
    32 °C
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    10.6 (5 g/l, H2O, 20 °C)
  • Ignition Point:
    275 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specificaties

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specificaties