Fmoc-Ile-Oh 1 * 100 g
Catalog # 04-12-1024-100
Supplier: Novabiochem (Part of Merck)
Frequently Bought Together
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Cell Strainers
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection