Order Entry
Netherlands
Orders LinkContactUsLinkComponent
D(+)-Maltose

D(+)-Maltose

Glass bottle for solids, 250 g
Catalog # SERA28390.02
Marketsource
Supplier:   Serva
D(+)-Maltose
Catalog # SERA28390.02
Supplier:  Serva
Marketsource
CAS Number:  69-79-4
D(+)-Maltose

Glass bottle for solids, 250 g

CAS Number:  69-79-4
Price
49,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, Maltose
  • CAS Number:69-79-4
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00135877
  • Molecular Weight:342.3 g/mol
  • Merck Index:12,05753
  • Melting Pt:494 °C
  • Density:1.532 g/cm³ (20 °C)
  •  :Density: 1.532 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-716-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.