Crystal violet
Glass bottle for solids, 100 g
Catalog # SERA27335.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Melting Pt:189…194 °C
- Density:1.19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:208-953-6
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



