Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Hydroxyurea

Hydroxyurea

Glass bottle for solids, 1 g
Catalog # SERA25435.01
Marketsource
Supplier:   Serva
Hydroxyurea
Catalog # SERA25435.01
Supplier:  Serva
Marketsource
CAS Number:  127-07-1
Hydroxyurea

Glass bottle for solids, 1 g

CAS Number:  127-07-1
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…144 °C
  • Storage Temperature:Ambient
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.