Order Entry
Netherlands
Orders LinkContactUsLinkComponent
DEAE-Dextran

DEAE-Dextran

Glass bottle for solids, 25 g
Catalog # SERA18686.01
Marketsource
Supplier:   Serva
DEAE-Dextran
Catalog # SERA18686.01
Supplier:  Serva
Marketsource
CAS Number:  9015-73-0
DEAE-Dextran

Glass bottle for solids, 25 g

CAS Number:  9015-73-0
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Diethylaminoethyl dextran
  • CAS Number:9015-73-0
  • MDL Number:MFCD00081561
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications