Dexamethasone
Glass bottle for solids, 100 mg
SERA18660.01
Marketsource
: Serva
: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
- :50-02-2
- :C₂₂H₂₉FO₅
- :MFCD00064136
- :392.47 g/mol
- :268…271 °C
- :200-003-9
- Pack type:Glass bottle for solids
- Pk:100 mg