Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sodium cholate

Sodium cholate

Glass bottle for solids, 100 g
Catalog # SERA17126.02
Marketsource
Supplier:   Serva
Sodium cholate
Catalog # SERA17126.02
Supplier:  Serva
Marketsource
CAS Number:  361-09-1
Sodium cholate

Glass bottle for solids, 100 g

CAS Number:  361-09-1
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt
  • CAS Number:361-09-1
  • Formula:C₂₄H₃₉NaO₅
  • MDL Number:MFCD00150749
  • Molecular Weight:430.56 g/mol
  • Melting Pt:198 °C
  • Storage Temperature:Ambient
  • EINECS:206-643-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.