Order Entry
Netherlands
Orders LinkContactUsLinkComponent
D-(+)-Cellobiose

D-(+)-Cellobiose

Glass bottle for solids, 50 g
Catalog # SERA16400.02
Marketsource
Supplier:   Serva
D-(+)-Cellobiose
Catalog # SERA16400.02
Supplier:  Serva
Marketsource
CAS Number:  528-50-7
D-(+)-Cellobiose

Glass bottle for solids, 50 g

CAS Number:  528-50-7
Price
Inquire For Price
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS Number:528-50-7
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00136034
  • Molecular Weight:342.30 g/mol
  • Storage Temperature:Ambient
  • EINECS:208-436-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications