Anthrone
Glass bottle for solids, 50 g
Catalog # SERA13680.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- EINECS:201-994-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



