Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Anthrone

Anthrone

Glass bottle for solids, 10 g
Catalog # SERA13680.01
Irritant
Marketsource
Supplier:   Serva
Anthrone
Catalog # SERA13680.01
Supplier:  Serva
Marketsource
CAS Number:  90-44-8
Anthrone

Glass bottle for solids, 10 g

CAS Number:  90-44-8
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • EINECS:201-994-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.