Order Entry
Netherlands
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

20 g, 2,5 ± mg
Catalogus # 611-8285
Leverancier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalogus # 611-8285
Leverancier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

20 g, 2,5 ± mg

Price
55,70 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specificaties
  • Weight:
    20 g
  • Seq:
    14
  • Tolerance:
    2,5 ± mg
Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.