Order Entry
Netherlands
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

10 g, 2,0 ± mg
Catalogus # 611-8284
Leverancier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalogus # 611-8284
Leverancier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

10 g, 2,0 ± mg

Price
55,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specificaties
  • Weight:
    10 g
  • Seq:
    13
  • Tolerance:
    2,0 ± mg