Order Entry
Netherlands
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1

VWR®, Calibration Weights, Class M1

2 g, 1,2 ± mg
Catalogus # 611-8282
Leverancier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalogus # 611-8282
Leverancier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1

2 g, 1,2 ± mg

Price
52,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.

Delivery information: All weights are supplied in plastic cases.

Specificaties
  • Weight:
    2 g
  • Seq:
    11
  • Tolerance:
    1,2 ± mg