VWR®, Calibration Weights, Class M1
1 g, 1,0 ± mg
611-8281
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.
- Weight:1 g
- Seq:10
- Tolerance:1,0 ± mg
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
: All weights are supplied in plastic cases.