Order Entry
Netherlands
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
2 mg, 0,2 ± mg
Catalogus # 611-8273
Leverancier:   VWR Collection
VWR®, Calibration Weights, Class M1
Catalogus # 611-8273
Leverancier:  VWR Collection
VWR®, Calibration Weights, Class M1
VWR®, Calibration Weights, Class M1
2 mg, 0,2 ± mg
Price
48,10 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Specificaties
  • Weight:
    2 mg
  • Seq:
    02
  • Tolerance:
    0,2 ± mg
Aanbevelingen