VWR®, Calibration Weights, Class M1
50 g, 3,0 ± mg
Catalogus # 611-8264
Leverancier: VWR Collection
Ga naar:
Product Details & Documents
Weights from 1 to 50 g are monobloc weights, with larger weights having an adjusting cavity.
Delivery information: All weights are supplied in plastic cases.
Documents
Specificaties
- Weight:50 g
- Seq:40
- Tolerance:3,0 ± mg