Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Funnels, analytical, DURAN®

Funnels, analytical, DURAN®

150 mm, 9 mm, 65 mm
Catalog # 221-1711
Supplier:   DWK Life Sciences
Funnels, analytical, DURAN®
Catalog # 221-1711
Supplier:  DWK Life Sciences
Funnels, analytical, DURAN®
Funnels, analytical, DURAN®

150 mm, 9 mm, 65 mm

Price
29,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

For rapid filtration.

Specifications
  • Funnel O-Ø:
    65 mm
  • Stem O-Ø:
    9 mm
  • Stem length:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.