Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS, -
Catalog # 527-1324
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # 527-1324
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
500 ml, 29/32 NS, -
Price
78,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicaatglas, met NS-PE-stop.
Met socket en gewone steel. Beschikbaar met vervangbare volglazen-kraanplug of PTFE-kraanplug voor vetvrije werking. Beiden met of zonder graduatie.
Documents
Specifications
  • Huls:
    29/32 NS
  • Capaciteit:
    500 ml
  • Indeling:
    -