AGITATEURS ROTATIFS, ROTARY MIXER, LABINCO, AXES POUR ROTARY MIXER, POUR: L26/27, LD76, TUBES Ø (MM): 12 - 16, LONGUEUR MAX. (MM): 1 20, NBRE DE TUBES: 48 1 * 1 Pce
Catalog # LBNC26480
Supplier: LABINCO
AGITATEURS ROTATIFS, ROTARY MIXER, LABINCO, AXES POUR ROTARY MIXER, POUR: L26/27, LD76, TUBES Ø (MM): 12 - 16, LONGUEUR MAX. (MM): 1 20, NBRE DE TUBES: 48 1 * 1 Pce
Catalog # LBNC26480
Supplier: LABINCO
Frequently Bought Together
Magnetic hotplate stirrers, ARE/AREX 5 series
Supplier: VELP SCIENTIFIC
Accessories and stirring tools for ES, PW, BS, DLS, LH and DLH stirrers
Supplier: VELP SCIENTIFIC
VWR® Professional 3500, Incubating Orbital Shaker
Supplier: VWR Collection
Magnetic stirrer, MIX 1 XL
Supplier: 2MAG
Accessories for shakers
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Disruptor Beads™ for Disruptor Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories for VWR® Advanced and Professional Hotplates and Hotplate Stirrers
Supplier: VWR Collection
TAP FOR MICROLITER SYRINGE 1 * 1 items
Supplier: Bohlender
250ml square media bottle clamp for Solaris Shaker Platforms 1 * 1 items
Supplier: Thermo Fisher Scientific
PressureSyn Glass Liner 1 * 1 items
Supplier: ASYNT
Multi-position magnetic stirrers, MIXdrive series
Supplier: 2MAG
AM 4 Digital Multi-position Hotplate Stirrers
Supplier: VELP SCIENTIFIC
Undergrad analogue magnetic stirrers, Stuart®
Supplier: Cole Parmer
Flask Stand & Clamp Kit 1 * 1 items
Supplier: Heidolph Instruments GmbH & Co.KG
Stirring blades, moon-shaped
Supplier: Bohlender