Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Pipette tips, gel loading, ART™
Pipette tips, gel loading, ART™
ART 100 GEL, 100 µl, +, +, 68,6 mm, 10 racks, 96 each
Catalog # 732-2226
   Low Manufacturing Impact
Pipette tips, gel loading, ART™
Catalog # 732-2226
   Low Manufacturing Impact
Pipette tips, gel loading, ART™
Pipette tips, gel loading, ART™
ART 100 GEL, 100 µl, +, +, 68,6 mm, 10 racks, 96 each
Price
96,00 €
Sale
Valid until 31/03/2026
Log in for availability
1 * 960 ST
Product Details & Documents
These tips are designed for loading vertical acrylamide or agarose gels.
  • Sterile tips are certified free from RNase, DNase, ATP and pyrogen
  • Available with or without filter for trapping potential contaminants
  • Manufactured according to the industry's highest quality standard
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    100 µl
  • Length:
    68,6 mm
  • Seq:
    03
  • Tip style:
    ART 100 GEL
  • Filter:
    +
  • Cat. No.:
    732-2226
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact