Boranepyridine complex 95%
Glass bottle, 25 mL
: Pyridine Borane, Borane, pyridine complex
- :110-51-0
- :C₅H₈BN
- :MFCD00012435
- :92.94 g/mol
- :10…11 °C
- :0.92 g/cm³
- :73 °C
- :Refrigerator
- :2810
- :6.1,II
Documents
ACRO17701
SDS
- Appearance:Clear to slightly hazy colorless to amber Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Titration:≥94 % (Active hydrogen)
- HPLC:≥95.0 %
- Pack type:Glass bottle
- Pk:25 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



