Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Morpholine 99+%, extra pure

Glass bottle, 2,5 L
  ACRO158680025
 
 
 
Morpholine 99+%, extra pure
  ACRO158680025
 :  110-91-8
Morpholine 99+%, extra pure
Morpholine 99+%, extra pure

Glass bottle, 2,5 L

 :  110-91-8
 
67,40 €
   
1 * 2,5 L
 
  •  :110-91-8
  •  :C₄H₉NO
  •  :MFCD00005972
  •  :87.12 g/mol
  •  :12,06362
  •  :126…130 °C (760 mmHg)
  •  :–5 °C
  •  :0.99 g/cm³
  •  :32 °C
  •  :Ambient
  •  :203-815-1
  •  :2054
  •  :8,I

 

 

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4550 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤20 APHA
  • Insoluble matter:
    ≤0.01 %

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L