Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Morpholine 99+%, extra pure

Glass bottle, 1 L
Catalog # ACRO158680010
Toxic
Corrosive
Flammable
Morpholine 99+%, extra pure
Catalog # ACRO158680010
CAS Number:  110-91-8
Morpholine 99+%, extra pure
Morpholine 99+%, extra pure

Glass bottle, 1 L

CAS Number:  110-91-8
Price
51,40 €
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:12,06362
  • Boiling Pt:126…130 °C (760 mmHg)
  • Melting Pt:–5 °C
  • Density:0.99 g/cm³
  • Flash Pt:32 °C
  • Storage Temperature:Ambient
  • EINECS:203-815-1
  • UN:2054
  • ADR:8,I

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4530 to 1.4550 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤20 APHA
  • Insoluble matter:
    ≤0.01 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications