Propionic anhydride 99%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Propionic anhydride 99%
Catalog # ACRO131520010
Supplier: Thermo Fisher Scientific
CAS Number: 123-62-6
Jump to:
Product Details & Documents
- CAS Number:123-62-6
- Formula:C₆H₁₀O₃
- MDL Number:MFCD00009303
- Molecular Weight:130.14 g/mol
- Merck Index:12,08011
- Boiling Pt:167 °C (1013 hPa)
- Melting Pt:–42 °C
- Density:1.015 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Ambient
- EINECS:204-638-2
- UN:2496
- ADR:8,III
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4030 to 1.4050 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



