Order Entry
Netherlands
ContactUsLinkComponent
IL-6, Rat, Biotrak Assay, ELISA - 96 wells (Range: 31-2000 pg/ml. Sensitivity: <16 pg/ml. Sample volume: 50ul. Sensitivity: 0.1ng/ml . 1.5L of wash buffer supplied) 1 * 96 Assays
Catalog # RPN2742
Supplier:  Cytiva
IL-6, Rat, Biotrak Assay, ELISA - 96 wells (Range: 31-2000 pg/ml. Sensitivity: <16 pg/ml. Sample volume: 50ul. Sensitivity: 0.1ng/ml . 1.5L of wash buffer supplied) 1 * 96 Assays
Catalog # RPN2742
Supplier:  Cytiva
Frequently Bought Together