Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Melatonin 99%

Melatonin 99%

Glass bottle, 250 mg
Catalog # ACRO125362500
Melatonin 99%
Catalog # ACRO125362500
CAS Number:  73-31-4
Melatonin 99%
Melatonin 99%

Glass bottle, 250 mg

CAS Number:  73-31-4
Price
49,60 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • Boiling Pt:512.8 °C
  • Melting Pt:118 °C
  • Flash Pt:264 °C
  • Storage Temperature:Ambient
  • EINECS:200-797-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    116 to 120 °C
  • Loss on drying:
    ≤0.5 % (1 g, 105 °C)
  • Heavy metals:
    ≤20 ppm
  • HPLC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.