L(+)-Glutamine 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
- CAS Number:56-85-9
- Formula:C₅H₁₀N₂O₃
- MDL Number:MFCD00008044
- Molecular Weight:146.15 g/mol
- Merck Index:13,04484
- Boiling Pt:355 °C (1013 hPa)
- Melting Pt:185 °C (dec.)
- Density:1,5326 g/cm³ (130 °C)
- Storage Temperature:Ambient
- EINECS:200-292-1
Documents
ACRO11995
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 % (105°C, 3 h)
- Sulfated ash:≤0.3 %
- Specific optical rotation:+31.5° to +33.0° (20°C, 589 nm) (c=10, 2N HCl)
- Titration with HClO4:98.5 to 101.5 %
- Iron (Fe):≤30 ppm
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



