Ethyl bromopyruvate 80-85%
Glass bottle, 25 mL
: Bromopyruvic acid ethyl ester, Ethyl 3-bromo-2-oxopropanoate, 3-Bromo-2-oxopropionic acid ethyl ester
- :70-23-5
- :BrH₂CC(O)COOCH₂CH₃
- :MFCD00000204
- :195.01 g/mol
- :100…102 °C (15 mmHg)
- :1.55 g/cm³
- :98 °C
- :Refrigerator
- :200-729-6
- :3265
- :8,III
- :Density: 1.55 g/cm³ (20 °C)
- Appearance:Clear yellow to pink-red Liquid
- Infrared spectrum:Conforms
- GC:≥80 %
- Phosphate (PO4):≤10 ppm
- Sulfate (SO4):≤30 ppm
- Sodium carbonate (Na2CO3):≤1.0 %
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤20 ppm
- Mercury (Hg):≤0.1 ppm
- Nickel (Ni):≤10 ppm
- Potassium (K):≤200 ppm
- Heavy metals (as Ag):≤20 ppm
- Nitrogen compounds (as N):≤0.001 %
- Pack type:Glass bottle
- Pk:25 mL



