Sodium deoxycholate, Millipore®
Glass bottle for solids, 1 kg
Supplier: Merck Millipore (Calbiochem)
Sodium deoxycholate, Millipore®
Catalog # 264101-1
Supplier: Merck Millipore (Calbiochem)
CAS Number: 302-95-4
Jump to:
Product Details & Documents
Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
- CAS Number:302-95-4
- Formula:C₂₄H₃₉NaO₄
- MDL Number:MFCD00064139
- Molecular Weight:414.56 g/mol
- Melting Pt:357…365 °C
- Storage Temperature:Ambient
- EINECS:206-132-7
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



