Thermometer, Landsberger, mercury filled, measuring range: 67:85/0.05C, enclosed scale, capillary lens front, colourless, total im m ersion, dimension: 300x9mm, richter-top finish with button, suitable for government verification. Until 10th Octobe 1 * 1 items
Catalog # AMARN63808
Supplier: Amarell
Frequently Bought Together