Order Entry
Netherlands
ContactUsLinkComponent
Thermometer, Anschutz milk glass type, mercury filled, measuring range:-48:102/0.1C, enclosed scale, capillary lens front, colourl e ss, dimension: 340x5-6mm, Richter-top finish, government verified with verification certificate. Until 10th October 1 * 1 items
Catalog # AMARN60143
Supplier:  Amarell
Thermometer, Anschutz milk glass type, mercury filled, measuring range:-48:102/0.1C, enclosed scale, capillary lens front, colourl e ss, dimension: 340x5-6mm, Richter-top finish, government verified with verification certificate. Until 10th October 1 * 1 items
Catalog # AMARN60143
Supplier:  Amarell
Frequently Bought Together