Alcoholometer, mercury filled, EUROPA classII, measuring range:80-90:0.1%vol, with thermometer, ranging:5:25/0.2C, length:400mm, s u itable for government verification. Until 10th October 2017 it is still allowed to use mercury thermometers if they 1 * 1 items
Catalog # AMARH848176S
Supplier: Amarell
Frequently Bought Together