Order Entry
Netherlands
ContactUsLinkComponent
Range finder, set, DIN12793, measuring range:0.60-2.00:0.005g/cm , without thermometer, 4 instruments in stuffed case, reference tem perature:20°C. 1 * 1 items
Catalog # AMARH820490
Supplier:  Amarell
Range finder, set, DIN12793, measuring range:0.60-2.00:0.005g/cm , without thermometer, 4 instruments in stuffed case, reference tem perature:20°C. 1 * 1 items
Catalog # AMARH820490
Supplier:  Amarell
Frequently Bought Together