Order Entry
Netherlands
ContactUsLinkComponent
Thermometer, maximum, for swinging, mercury filled, measuring range:-10:250/1C, length:300mm, diameter:7-8mm, colourless, capillar y lens front, enclosed scale, with ring at top. Until 10th October 2017 it is still allowed to use mercury thermomete 1 * 1 items
Catalog # 620-1104
Supplier:  Amarell
undefined
Thermometer, maximum, for swinging, mercury filled, measuring range:-10:250/1C, length:300mm, diameter:7-8mm, colourless, capillar y lens front, enclosed scale, with ring at top. Until 10th October 2017 it is still allowed to use mercury thermomete 1 * 1 items
Catalog # 620-1104
Supplier:  Amarell
Frequently Bought Together