Streptavidin
Plastic bottle for solids, 10 mg
Catalog # 189730-10
Supplier: Merck Millipore (Oncogene)
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:10 mg
- Cat. No.:189730-10
- Protein/peptide name:Streptavidin
- Storage conditions:Freezer
- MDL No.:MFCD00082035
- CAS No.:9013-20-1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



